missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxJ1/HFH4 Antibody (CL3991) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59033FR
This item is not returnable.
View return policy
Description
FoxJ1/HFH4 Monoclonal antibody specifically detects FoxJ1/HFH4 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| FoxJ1/HFH4 | |
| Monoclonal | |
| DyLight 680 | |
| FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202 | |
| This antibody was developed using a recombinant protein derived from Q92949, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 2302 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3991 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction