Learn More
FoxJ1/HFH4 Antibody (CL3989) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59032CL1
Product Code. 30290935
  • 1175.00€ / 0.10mL
Description
FoxJ1/HFH4 Monoclonal antibody specifically detects FoxJ1/HFH4 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FoxJ1/HFH4
Monoclonal
CoraFluor 1
FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202
This antibody was developed using a recombinant protein derived from Q92949, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Transcription Factors and Regulators
2302
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL3989
PBS
Mouse
Protein A purified