missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXH1 Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5148822

Product Code. 30535606

  • 499.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXH1
Polyclonal
Unconjugated
FOXH1
contains winged-helix domain; etID62177.12; fast; Fast1; Fast-1; FAST1 transcription factor; FAST2; fast-2; fe26b06; Forkhead activin signal transducer 1; forkhead activin signal transducer 2; forkhead activin signal transducer FAST; forkhead activin signal transducer-1; forkhead box H1; forkhead box protein H1; Foxh1; hFAST-1; schmalspur; Schmalspur protein; SUR; TGF-beta/activin signal transducer; transcription factor; uncle freddy; unf; wu:fe26b06; wu:fi30c08; wu:fi32b01
Sheep
Antigen affinity chromatography
RUO
8928
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with trehalose and no preservative
O75593
FOXH1
E. coli-derived recombinant human FoxH1, Ser164-Ser272.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.