missing translation for 'onlineSavingsMsg'
Learn More

FOXE1 Antibody, Novus Biologicals™

Product Code. 18250000 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18250000 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18250000 Supplier Novus Biologicals Supplier No. NBP130960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXE1 Polyclonal specifically detects FOXE1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXE1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Alias FKHL15HFKL5, forkhead box E1 (thyroid transcription factor 2), forkhead box E2, Forkhead box protein E2, forkhead, drosophila, homolog-like 15, Forkhead-related protein FKHL15, HFKH4Thyroid transcription factor 2, TITF2HNF-3/fork head-like protein 5, TTF2forkhead box protein E1, TTF-2FOXE2
Gene Symbols FOXE1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXE1. The exact sequence is proprietary.
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2304
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%) and Rat (85%). Not yet tested in any other species.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.