missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXD3 Monoclonal Antibody (800602)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561517
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
Specifications
| FOXD3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q61060 | |
| FOXD3 | |
| E. coli-derived recombinant mouse FoxD3, Met1-Val130. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 800602 | |
| Unconjugated | |
| FOXD3 | |
| AIS1; CWH3; forkhead box D3; forkhead box protein D3; FOXD3; Genesis; hepatocyte nuclear factor 3 forkhead homolog 2; HFH2; HFH-2; HNF3/FH transcription factor genesis; HNF-3/forkhead homolog 2; VAMAS2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 15221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction