Learn More
Four elements solution - Ca, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
Product Code. p-7083773 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
Product Code. 12996193
Brand: Fisher Chemical MS 010 050
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Chemical Name or Material Four Elements
Packaging Bottle
Analyte or Component Names Ca,Mg,K,Na
Physical Form Liquid
Concentration 1000 mg/L
Type ICP/MS Calibration Multi-element Standard Solution
Element Ca, Mg, K, Na
Solvent or Matrix 2% HNO3
Quantity 50 mL