missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FosB Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148574
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
Specifications
| FosB | |
| Polyclonal | |
| Unconjugated | |
| Fosb | |
| activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 2354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P53539 | |
| Fosb | |
| E. coli-derived recombinant human FosB, Met1-Leu338. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction