missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formoterol EP Impurity G (as Hydrochloride), Mikromol
Description
Formoterol EP Impurity G (as Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | 4-Methoxyamphetamine (hydrochloride) |
| CAS | 3706-26-1 |
| Analyte or Component Names | 4-Methoxyamphetamine (hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-1-(4-Methoxyphenyl)propan-2-amine Hydrochloride, Formoterol Fumarate Dihydrate Imp. G (EP), Formoterol Imp. G (EP), (2RS)-1-(4-Methoxyphenyl)propan-2-amine Hydrochloride, Formoterol Fumarate Dihydrate Imp. G (EP) as Hydrochloride, PMA HCl, p-Methoxyamphetamine Hydrochloride, Formoterol Related Compound G as Hydrochloride, Formoterol Fumarate Dihydrate Impurity G as Hydrochloride, Formoterol Impurity G as Hydrochloride |
| InChI Formula | InChI=1S/C10H15NO.ClH/c1-8(11)7-9-3-5-10(12-2)6-4-9;/h3-6,8H,7,11H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.COc1ccc(CC(C)N)cc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?