missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR1 Antibody (2G5C12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00€ - 583.00€
Specifications
| Antigen | FOLR1 |
|---|---|
| Clone | 2G5C12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075934
|
Novus Biologicals
NBP2-61773 |
0.1 mL |
583.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660689
|
Novus Biologicals
NBP2-61773-0.025ml |
0.025 mL |
196.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOLR1 Monoclonal specifically detects FOLR1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifica
| FOLR1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Cancer | |
| 2348 | |
| Purified recombinant fragment of human FOLR1 (AA: 41-227) expressed in E. Coli. | |
| Primary | |
| 29.8 kDa |
| 2G5C12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18 | |
| FOLR1 | |
| IgG2a | |
| Protein G purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto