missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FMR1 (Human) Recombinant Protein

Human FMR1 full-length ORF ( AAH38998.1, 1 a.a. - 297 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002332-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195081

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MEELVVEVRGSNGAFYKAFVKDVHEDSITVAFENNWQPDRQIPFHDVRFPPPVGYNKDINESDEVEVYSRANEKEPCCWWLAKVRMIKGEFYVIEYAACDATYNEIVTIERLRSVNPNKPATKDTFHKIKLDVPEDLRQMCAKEAAHKDFKKAVGAFSVTYDPENYQLVILSINEVTSKRAHMLIDMHFRSLRTKLSLIMRNEEASKQLESSRQLASRFHEQFIVREDLMGLAIGTHGANIQQARKVPGVTAIDLDEDTCTFHIYGEDQDAVKKARSFLEFAEDVIQVPRNLVGLKI
Specifications

Specifications

AAH38998.1
fragile X mental retardation 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FMR1
GST
2332
Wheat germ expression system
10 μg
FMRP, FRAXA, MGC87458, POF, POF1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers