missing translation for 'onlineSavingsMsg'
Learn More

FMOC-D-Beta-homoalanine, TRC

Product Code. 30178952
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178952 1 g 1g
30179547 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30178952 Supplier Toronto Research Chemicals Supplier No. TRCF6576731G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201864-71-3
Molecular Formula C19 H19 N O4
Molecular Weight (g/mol) 325.36
Synonym Butanoic acid, 3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-, (R)-, (3R)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]butanoic acid, (3R)-3-([[(9H-Fluoren-9-yl)methoxy]carbonyl]amino)butanoic acid
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C[C@H](CC(=O)O)NC(=O)OCC1c2ccccc2c3ccccc13

Specifications

InChI Formula InChI=1S/C19H19NO4/c1-12(10-18(21)22)20-19(23)24-11-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,12,17H,10-11H2,1H3,(H,20,23)(H,21,22)/t12-/m1/s1
Quantity 1 g
Formula Weight 325.1314
Shipping Condition Room Temperature
Chemical Name or Material FMOC-D-beta-Homoalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.