CAS RN 201864-71-3
CAS RN 201864-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
FMOC-D-Beta-homoalanine, TRC
CAS: 201864-71-3 Molecular Formula: C19 H19 N O4 Molecular Weight (g/mol): 325.36 Synonym: Butanoic acid, 3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-, (R)-,(3R)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]butanoic acid,(3R)-3-([[(9H-Fluoren-9-yl)methoxy]carbonyl]amino)butanoic acid IUPAC Name: (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid SMILES: C[C@H](CC(=O)O)NC(=O)OCC1c2ccccc2c3ccccc13