Learn More
FMOC-D-Beta-homoalanine, TRC
Product Code. 30179547
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179547 500 mg 500mg
30178952 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179547 Supplier Toronto Research Chemicals Supplier No. TRCF657673500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 201864-71-3
Molecular Formula C19 H19 N O4
Molecular Weight (g/mol) 325.36
Synonym Butanoic acid, 3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-, (R)-, (3R)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]butanoic acid, (3R)-3-([[(9H-Fluoren-9-yl)methoxy]carbonyl]amino)butanoic acid
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C[C@H](CC(=O)O)NC(=O)OCC1c2ccccc2c3ccccc13
Specifications
InChI Formula InChI=1S/C19H19NO4/c1-12(10-18(21)22)20-19(23)24-11-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,12,17H,10-11H2,1H3,(H,20,23)(H,21,22)/t12-/m1/s1
Quantity 500 mg
Formula Weight 325.1314
Shipping Condition Room Temperature
Chemical Name or Material FMOC-D-beta-Homoalanine