Learn More
Fluoxetine EP Impurity B (as Hydrochloride), Mikromol
Product Code. 30460091
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460091 25 mg 25mg
30459947 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30460091 Supplier Mikromol Supplier No. MM0256.020025
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fluoxetine EP Impurity B (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1M-3PP HCl (Methyl-3-phenylpropylamine Hydrochloride)
CAS 30684-07-2
Analyte or Component Names 1M-3PP HCl (Methyl-3-phenylpropylamine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C10 H15 N . Cl H
Certifications/Compliance ISO 17025
Synonym N-Methyl-3-phenylpropan-1-amine Hydrochloride (Methyl-3-phenylpropylamine Hydrochloride), Fluoxetine Hydrochloride Imp. B (EP), Fluoxetine Imp. B (EP), Fluoxetine USP Related Compound B, Fluoxetine USP RC B, N-Methyl-3-phenylpropan-1-amine Hydrochloride, Fluoxetine Hydrochloride Imp. B (EP) as Hydrochloride, Methyl-3-phenylpropylamine Hydrochloride, 1M-3PP HCl, Fluoxetine Related Compound B as Hydrochloride, Fluoxetine Hydrochloride Impurity B as Hydrochloride, Fluoxetine Impurity B as Hydrochloride
InChI Formula InChI=1S/C10H15N.ClH/c1-11-9-5-8-10-6-3-2-4-7-10;/h2-4,6-7,11H,5,8-9H2,1H3;1H
Solution Type Neat
SMILES Cl.CNCCCc1ccccc1
IUPAC Name N-methyl-3-phenylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 185.69
Quantity 25 mg
Formula Weight 185.0971 g/mol