missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone Acetate, Mikromol

Product Code. 30458491
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458491 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458491 Supplier Mikromol Supplier No. MM0870.010100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fluorometholone Acetate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorometholone Acetate
CAS 3801-06-7
Analyte or Component Names Fluorometholone Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H31 F O5
Certifications/Compliance ISO 9001
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17-dihydroxy-6α-methyl-, 17-acetate (6CI,7CI,8CI), (6α,11β)-17-(Acetyloxy)-9-fluoro-11-hydroxy-6-methylpregna-1,4-diene-3,20-dione, 6α-Methyl-9α-fluoro-17-acetoxy-21-deoxyprednisolone, 6α-Methyl-9α-fluoro-21-desoxyprednisolone-17-acetate, Eflone, Flarex, Fluorometholone acetate, NSC 47438, Oxylone acetate, U 17323
InChI Formula InChI=1S/C24H31FO5/c1-13-10-19-17-7-9-23(14(2)26,30-15(3)27)22(17,5)12-20(29)24(19,25)21(4)8-6-16(28)11-18(13)21/h6,8,11,13,17,19-20,29H,7,9-10,12H2,1-5H3/t13-,17-,19-,20-,21-,22-,23-,24-/m0/s1
Solution Type Neat
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(=O)C)(C(=O)C)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11-hydroxy-6,10,13-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 418.5
Quantity 100 mg
Formula Weight 418.2156 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.