Learn More
Fluoride Standard Solution (Ion ECON), Chem Lab™
Click to view available options
Quantity:
500 mL
Specifications
Quantity | 500 mL |
Color | Colorless |
Odor | Odorless |
pH | ±9 |
Solubility | Soluble in water |
Physical Form | Liquid |