missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Pure, C.I.45350, Fisher Chemical™

Product Code. p-7073024 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10497250 25 g 25g
10598990 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10497250 Supplier Fisher Chemical Supplier No. F/1200/46

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number 5050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 314°C
Formula Weight 332.3g/mol
Packaging Glass Jar
Color Brown/Red
Quantity 25 g
Physical Form Powder
compliance-icons
Product Identifier
  • Fluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.