missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunisolide Hemihydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Flunisolide Hemihydrate
Specifications
Specifications
| Chemical Name or Material | Flunisolide Hemihydrate |
| CAS | 77326-96-6 |
| Analyte or Component Names | Flunisolide Hemihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C24 H31 F O6 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Flunisolide Hemihydrate, (6alpha,11beta,16alpha)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione Hydrate (2:1) |
| InChI Formula | InChI=1S/2C24H31FO6.H2O/c2*1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26;/h2*5-7,13-14,16-17,19-20,26,28H,8-11H2,1-4H3;1H2/t2*13-,14-,16-,17-,19+,20+,22-,23-,24+;/m00./s1 |
| SMILES | O.CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO.CC6(C)O[C@@H]7C[C@H]8[C@@H]9C[C@H](F)C%10=CC(=O)C=C[C@]%10(C)[C@H]9[C@@H](O)C[C@]8(C)[C@@]7(O6)C(=O)CO |
| IUPAC Name | (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione hemihydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction