missing translation for 'onlineSavingsMsg'
Learn More

Fludarabine, Mikromol™

Product Code. 30050826
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050826

Brand: Mikromol MM3428.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fludarabine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fludarabine
CAS 21679-14-1
Analyte or Component Names Fludarabine
Shipping Condition Room Temperature
Molecular Formula C10 H12 F N5 O4
Certifications/Compliance ISO 9001
Synonym 9H-Purin-6-amine, 9-beta-D-arabinofuranosyl-2-fluoro-, Adenine, 9-beta-D-arabinofuranosyl-2-fluoro- (8CI), 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 2-Fluoro Ara-A, 2-Fluoro-9-beta-D-arabinofuranosyladenine, 2-Fluoroadenine arabinoside, 2F-Ara-A, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, 9-beta-D-Arabinosyl-2-fluoroadenine, F-ara-A, Fludarabine, NSC 118218, NSC 118218H, Fludarabine Phosphate Impurity E
InChI Formula InChI=1S/C10H12FN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6+,9-/m1/s1
SMILES Nc1nc(F)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O
IUPAC Name (2R,3S,4S,5R)-2-(6-amino-2-fluoropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 285.23
Quantity 25 mg
Formula Weight 285.0873 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.