missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT4 Monoclonal Antibody (OTI8C2), TrueMAB™, OriGene
Description
FLT4 Monoclonal antibody specifically detects FLT4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | FLT4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8C2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | HMG20A |
| Gene Accession No. | Q9NP66 |
| Gene Alias | AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?