missing translation for 'onlineSavingsMsg'
Learn More

Flt-1/VEGFR1 Ab-1 Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in western blotting experiments with Epredia™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody.

183.00€ - 820.00€

Specifications

Antigen Flt-1/VEGFR1 Ab-1
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Formulation Price Quantity & Availability  
Product Code Brand Quantity Formulation Price Quantity & Availability  
12633037
View Documents
Epredia
RB-1527-P0
100 μL Purified with BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12643037
View Documents
Epredia
RB-1527-P1
500 μL Purified with BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12653037
View Documents
Epredia
RB-1527-P1ABX
500 μL Purified w/o BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12623037
View Documents
Epredia
RB-1527-P
1 mL Purified with BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
12663037
View Documents
Epredia
RB-1527-PABX
1 mL Purified w/o BSA and Azide This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Flt-1/vascular endothelial growth factor receptor 1 (VEGFR1), a cell membrane receptor kinase, is a high-affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flk-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin

Molecular Weight: 180 kDa

Positive Control: Hey cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Flt-1/VEGFR1 Ab-1
Western Blot
Unconjugated
Rabbit
Angiogenesis
A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin
Primary
1mg/mL
Polyclonal
Concentrated
RUO
Human, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.