missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flotillin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16508
This item is not returnable.
View return policy
Description
Flotillin-1 Polyclonal specifically detects Flotillin-1 in Human, Hamster samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Flotillin-1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7) with 0.01% Thimerosal | |
| flotillin 1, flotillin-1, integral membrane component of caveolae | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Monkey (100%), Pig (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Simple Western reported by internal validation, Immunoprecipitation 1:100-1:500 | |
| O75955 | |
| FLOT1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Flotillin 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10211 | |
| Human, Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction