missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Antibody (4E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15148
This item is not returnable.
View return policy
Description
FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| FLI1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 400000 | |
| Western Blot 1:500-1:5000, ELISA | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| A synthesized peptide derived from human FLI1. UniProt ID (Q01543). | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 2313 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction