Learn More
Flavoxate Hydrochloride, Mikromol™
Description
Flavoxate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Flavoxate Hydrochloride |
| CAS | 3717-88-2 |
| Analyte or Component Names | Flavoxate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H25 N O4 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Flavoxate Hydrochloride, 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid 2-(1-piperidinyl)ethyl ester hydrochloride, 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid 2-piperidinoethyl ester hydrochloride, 1-Piperidineethanol 3-methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (ester) hydrochloride, Bladderon, DW 61, Flavoxate hydrochloride, Genurin, NSC 114649, Patricin, Peflate, Piperidinoethyl-3-methylflavone-8-carboxylate hydrochloride, Rec 7/0040, Spasmal, Spasuret, Urispas |
| InChI Formula | InChI=1S/C24H25NO4.ClH/c1-17-21(26)19-11-8-12-20(23(19)29-22(17)18-9-4-2-5-10-18)24(27)28-16-15-25-13-6-3-7-14-25;/h2,4-5,8-12H,3,6-7,13-16H2,1H3;1H |
| SMILES | Cl.CC1=C(Oc2c(cccc2C1=O)C(=O)OCCN3CCCCC3)c4ccccc4 |
| IUPAC Name | 2-piperidin-1-ylethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate;hydrochloride |
| Show More |