Learn More
Agencinox™ Flat Bottom Stainless Steel Dishes
Product Code. p-8000005
Click to view available options
Capacity (Metric):
100 mL
250 mL
Unit Size:
Each
Product Code. 11756533
Brand: Agencinox™ 308
Only null left
Add to basket
Add to basket
Use these dishes in decontamination services, operating rooms, surgeries, clinics and healthcare fields. Agencinox™ Flat Bottom Stainless Steel Dishes are ideal for holding dressings and other items which may be needed during surgical procedures.
  • Flat bottom stainless steel lotion bowls
  • Autoclavable
  • Available in a variety of sizes
Specifications
Capacity (Metric) 100 mL
Material Stainless Steel (18/10)
Type Lotion Bowl
Height (Metric) 40 mm
Diameter (Metric) 80 mm