Learn More
Novus Biologicals™ FLAP Antibody, Novus Biologicals™
Product Code. 30066911 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066911 100 μg 100µg
30066924 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30066911 Supplier Novus Biologicals™ Supplier No. NBP330926100ug
Rabbit Polyclonal Antibody
FLAP Polyclonal antibody specifically detects FLAP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen FLAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1-5 μg/mL
Formulation Phosphate buffered saline
Gene Alias arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein
Host Species Rabbit
Immunogen Synthetic peptide derived from human FLAP protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 241
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified