Learn More
Idex Flangeless Ferrule, Blue ETFE, 1/16 in. OD Tubing, 1/4-28 Flat-Bottom
Click to view available options
Packaging Quantity:
10 Pk.
Unit Size:
Pack of 10
Description
- Excellent for connecting rigid or semi-rigid tubing.
- Use with male nuts for a leak-tight connection.
Specifications
Specifications
| Type | Fitting |
| Diameter (English) Outer | 0.0625 in. |
| Diameter (Metric) Outer | 1.5875 mm |
| Model | 0193930 |
| Packaging Quantity | 10 Pk. |
| Thread Size | 25.4 |