missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP15 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-424AF488
This item is not returnable.
View return policy
Description
FKBP15 Polyclonal antibody specifically detects FKBP15 in Human samples. It is validated for Western Blot
Specifications
| FKBP15 | |
| Polyclonal | |
| Western Blot | |
| FK506 binding protein 15, 133kDa, FK506-binding protein 133kDa, FK506-binding protein 15, FKBP133, FKBP-133, FKBP-15,133 kDa FKBP, PPP1R76,133 kDa FK506-binding protein, regulatory subunit 76, WAFL | |
| A synthetic peptide mapping to a region between residues 1175 and 1234 (C-terminus) of human FK506-binding protein 135 using the numbering given in TrEMBL entry Q9Y4D0 (GeneID 23307). | |
| 0.1 mL | |
| Signal Transduction | |
| 23307 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction