missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | FKBP12 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631306
|
Novus Biologicals
NBP2-99149-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631126
|
Novus Biologicals
NBP2-99149-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP12 Polyclonal antibody specifically detects FKBP12 in Human samples. It is validated for Western Blot, ELISASpecifications
| FKBP12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, mTOR Pathway | |
| 0.2 um filtered solution in PBS | |
| 2280 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Produced in rabbits immunized with purified, recombinant Human FKBP12 (Accession#: NP_463460; Met 1-Glu 108) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title