missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIZZ1/RELM alpha Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29355APC
This item is not returnable.
View return policy
Description
FIZZ1/RELM alpha Polyclonal antibody specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Flow Cytometry
Specifications
| FIZZ1/RELM alpha | |
| Polyclonal | |
| Flow Cytometry | |
| 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2 | |
| A synthetic peptide made to amino acids sequence within the residues 10-50 of the mouse RELM alpha protein. | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 57262 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction