missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Fixable Viability Dye, 750/777 Nm, 50 Tests
Description
- Assay type: Cell-based
- Applications: Fluorescence Microscopy, Flow Cyt
- Sample types: Adherent cells, Suspension cells
- Results type: Quantitative
- Reactive species: Mammals
- Protocol: Labeling in < 4 hrs with 30 secs hands-on time
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope |
| Format | Kit |
| Label or Dye | Fixable Viability Dye |
| No. of Reactions | 50 Tests |
| Reactivity | Mammals |
| Shipping Condition | Ambient |
| Target Molecule | Viable Cells |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?