Learn More
SCAT™ Square Installation Wrench for HPLC Systems
Product Code. 10510631 Shop All SCAT Europe Products
Click to view available options
Color:
Black
Unit Size:
Each
Product Code. 10510631
Brand: SCAT™ 160499
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
SCAT™ fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Type Wrench
Color Black
For Use With (Application) Solvent Handling
For Use With Polypropylene Fittings
Material PFA