Learn More
SCAT™ Fitting with Ferrule for HPLC Systems
Product Code. 10303970 Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Outer:
1.6 mm
3.2 mm
Color:
Black
Colorless
White
Unit Size:
Pack of 5
Product Code. 10303970
Brand: SCAT™ 107060
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
SCAT™ fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Type Fitting
Color Black
Diameter (Metric) Outer 3.2 mm
For Use With (Application) Solvent Handling
For Use With Capillary (3.2 mm OD)
Material PFA