Learn More
Traceable™ One-Shot Conductivity Standards, Fisherbrand™
Product Code. p-4321120 Shop All Fisherbrand Products
Click to view available options
Concentration:
150,000 μS/μ℧
200,000 μS/μ℧
Unit Size:
Pack of 6
Product Code. 11724256
Eliminates contamination with calibration within disposable container. Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.
  • Nominal Value: 150000 Micro (mhos/siemens)
  • Certified Reference Material
  • Accuracy: certificate stated uncertainty, or ±0.25 microsiemen or ±0.25%
  • Includes: 6-pack 100 mL container, step-by-step calibration instructions, individual temperature compensation charts, traceability information, Traceable Certificate
Specifications
Chemical Name or Material One-Shot Conductivity Standards
Packaging Bottle
CAS 7447-40-7
Molecular Formula ClK
Concentration 150,000 μS/μ℧
MDL Number MFCD00011360
Technique Calibration of Conductivity Instrument
Color Clear
For Use With (Equipment) Conductivity Meters and Probes
Solubility Soluble in water
Solution Type Calibration Solution
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Type Conductivity Standard
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.548
PubChem CID 4873
Quantity 6 x 100 mL
ChEBI CHEBI:32588