Learn More
Fisherbrand™ Mini-Centrifuge Replacement Parts
Replacement parts for Fisherbrand™ mini-centrifuge
65.45€
Specifications
Type | Fisherbrand™ Mini-centrifuge Replacement Parts |
---|---|
For Use With (Equipment) | Fisherbrand Mini-centrifuge |
Autoclavable | Not Autoclavable |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12883332
|
Fisherbrand™
|
Mini-centrifuge Rotor SnapSpin 6 x 1.5/2 mL Tube |
65.45€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
12893332
|
Fisherbrand™
|
Mini-centrifuge Rotor SnapSpin 2 x 8 Strip 0.2 mL |
65.45€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- For use with the Fisherbrand™ mini-centrifuge
Specifications
Fisherbrand™ Mini-centrifuge Replacement Parts | |
Fisherbrand Mini-centrifuge | |
Not Autoclavable |