Learn More
Fisherbrand™ SterilElite™ Tabletop Autoclaves
Product Code. 15525199 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
24 L
Depth (Metric) Outside:
660 mm
Voltage:
230 V
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Depth (Metric) Outside Voltage unitSize
15525199 24 L 660 mm 230 V 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15525199
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Combine compact design with high performance, reliability and safety.
Combine compact design with high performance, reliability and safety
Intelligent design for your professional choice.
  • Microprocessor control system provides nine fully automatic programs, including add water, sterilization and dry
  • Eight pre-installed sterilization cycles; custom cycles are not an option for the SterilElite16 or 24
  • Two separate water-level check mechanisms: water tank level and chamber water level
  • Auto dry function
  • Pressure door auto-lock device
  • Chamber protected by pressure switch and steam pressure safety valve
  • Vacuum/Pressure Release button cuts off the power supply, exhausts the chamber of pressure, and releases the chamber vacuum simultaneously
Indicator lights:
  • DOOR OPEN - Door has not been closed completely; all functions stop
  • LOW WATER - Water level is too low
  • COMPLETE - Full cycle has run properly and is complete
Order Info
Printer not included.
Specifications
Depth (English) Outside 26 in.
Depth (Metric) Outside 660 mm
Capacity (English) 25.4 qt.
Capacity (Metric) 24 L
Depth (Metric) Chamber 432 mm
Diameter (English) Chamber 10.23 in.
Depth (English) Chamber 17.0 in.
Diameter (Metric) Chamber 259.8 mm
For Use With (Application) Steam sterilization
Length (English) 26.0 in.
Height (English) Exterior 17.0 in.
Height (Metric) Exterior 440 mm
Length (Metric) 660 mm
Model Front-loading
Product Type Benchtop Autoclave
Temperature (Metric) 121°C to 135°C
Weight (Metric) 56 kg
Height (Metric) Chamber 228 mm
Height (English) Chamber 9 in.
Includes Eight pre-installed cycles
Frequency 50/60 Hz
Programmability Auto Dry
Voltage 230 V
Wattage 1800 w
Width (English) Exterior 21.25 in.
Width (Metric) Exterior 540 mm
Dimensions (L x W x H) 660 x 540 x 440 mm