Learn More
OHAUS™ Frontier™ 5306 Mini Centrifuge Accessory, Rotors
For use with Frontier 5306 Mini Centrifuge
Brand:  OHAUS™ 30129559
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15896612
  • 29.80€ / Each
Estimated Shipment: 07-06-2024
Specifications
Rotor, 1.5/2ml x8, FC5306
8 x 1.5/2 mL
6,000 rpm
45°
Rotor
38,800 x g
Frontier 5306 Mini Centrifuge
Angle rotor 8 x 1.5/2.0mL