missing translation for 'onlineSavingsMsg'
Learn More

Fipronil, Mikromol

Product Code. 30461617
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30461617

Brand: Mikromol MM0956.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fipronil, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fipronil
CAS 120068-37-3
Analyte or Component Names Fipronil
Shipping Condition Room Temperature
Molecular Formula C12 H4 Cl2 F6 N4 O S
Certifications/Compliance ISO 9001
Synonym Fipronil
InChI Formula InChI=1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2
Solution Type Neat
SMILES Nc1c(c(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylsulfinyl)pyrazole-3-carbonitrile
Molecular Weight (g/mol) 437.15
Quantity 250 mg
Formula Weight 435.9387 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.