missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1/RCP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | FIP1/RCP |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228477
|
Novus Biologicals
NBP3-38599-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232852
|
Novus Biologicals
NBP3-38599-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FIP1/RCP Polyclonal antibody specifically detects FIP1/RCP in Human samples. It is validated for ELISA,Western BlotSpecifications
| FIP1/RCP | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp686E2214, EC 3.2.1.28, EC 6.1.1.7, FLJ22524, FLJ22622, MGC78448, NOEL1A, RAB11 coupling protein, RAB11 family interacting protein 1 (class I), rab11 family-interacting protein 1, Rab11-FIP1, Rab-coupling protein, Rab-interacting recycling protein, RCPRab effector protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-540 of human FIP1/RCP (NP_079427.4).,, Sequence:, EAKGEIKDSSPSSSPSPKGFRKKHLFSSTENLAAGSWKEPAEGGGLSSDRQLSESSTKDSLKSMTLPSYRPAPLVSGDLRENMAPANSEATKEAKESKKPESRRSSLLSLMTGKKDVAKGSEGENPLTVPGREKEGMLMGVKPGEDASGPAEDLVRRSEKDTAAVVSRQGSSLNLFEDVQITEPEAEPESKSEPRPPISSPRAPQTRAVKP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 80223 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title