Learn More
Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips
GREENER_CHOICE
Product Code. p-4525583
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bagged
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 400
Pack of 6500
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Format unitSize
11408499 250 μL Non-sterile Bagged Pack of 1000
10053750 1000 μL Non-sterile Bagged Pack of 400
10329370 1000 μL Non-sterile Bulk Pack of 6500
10073560 1000 μL Sterile Rack Pack of 960
11428499 250 μL Non-sterile Rack Pack of 960
10349320 250 μL Sterile Rack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11408499 Supplier Thermo Scientific™ Supplier No. 9405020
Eliminate cell fragmentation caused by mechanical shearing with these wide orifice pipette tips, ideal for pipetting macromolecules.
Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, ideal for pipetting macromolecules.
Features:
  • Two different models cover volume range of 10-1000 μL
  • Wide orifice tip; inside diameter of orifice is 1.1mm/1.8mm
  • Color code: White
  • Length: 5.3cm/7.2cm
TRUSTED_SUSTAINABILITY
Specifications
Description Finntip 250 Wide, Non-Filtered, Non-sterile, Bulk
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Model Finntip 250 Wide
Length (English) 2 in.
Length (Metric) 5.3 cm
Format Bagged
Packaging 1000/Bag
Color Code White
For Use With (Equipment) Thermo Scientific Finnpipette 250
Product Line FinnTip Wide Orifice
Fit for Purpose Certified Quality