Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Product Code. 10115582
Click to view available options
Volume (Metric):
10 μL
250 μL
1000 μL
Unit Size:
Pack of 100
Pack of 50
Product Code. 10115582
Brand: Thermo Scientific™ 94053100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Perform PCR and other purity demanding applications with these biocon pipette tips, certified free from endotoxin, DNA, DNase and RNase.
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:
  • Individually packed
  • Three models in volume range 0.2-1000 μL
  • Presterilized with gamma radiation
  • Includes certificate of quality
Recommended for: PCR applications, molecular biology
TRUSTED_SUSTAINABILITY
Specifications
Description Finntip BioCon 10, Non-Filtered, Sterile,
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Model Finntip BioCon 10
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
For Use With (Equipment) Thermo Scientific FinnTip BioCon 10
Product Line FinnTip Biocon