Learn More
Thermo Scientific™ Finger Tite Connectors
Product Code. 10321485 Shop All Thermo Scientific Products
Click to view available options
Type:
Female Nut
Split/Splitless and FID Starter Kit
Split/Splitless and MS Starter Kit
Unit Size:
Each
Pack of 5
Product Code. 10321485
Brand: Thermo Scientific™ 290ST132
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure reliable, leak-free connections for optimal GC performance using these Finger Tite Connectors.
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
Recommended Use
  • Thermo Scientific™ and Agilent™ GC instruments
  • Connectors for SSL and MS systems
TRUSTED_SUSTAINABILITY
Specifications
Type Split/Splitless and FID Starter Kit
Description Injector/Detector Starter Kit
For Use With (Equipment) TRACE GC Ultra and FOCUS GC Instruments