Learn More
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Shop All Thermo Scientific Products
Click to view available options
Type:
Split/Splitless Injector Base Seal
Split/Splitless and FID Starter Kit
Split/Splitless and MS Starter Kit
Unit Size:
Each
Pack of 2
Descripción
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- Easy to handle with the nut touchable even with a hot injector/detector
- No tools required
Especificaciones
Especificaciones
| Type | Split/Splitless Injector Base Seal |
| Description | Injector Base Seal; 2/Pk. |
| For Use With (Equipment) | TRACE 1300 Series GC and Agilent GC Instruments |
| Quantity | 2/Pk. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido