Learn More
Finasteride, Thermo Scientific™
Inhibitor of steroid 5-a-reductase
Brand:  Thermo Scientific Alfa Aesar J63454.MC
Quantity 100mg
 View more versions of this product
Product Code. 15455699
  • 61.80€ / 100mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
98319-26-7
372.553
DBEPLOCGEIEOCV-WSBQPABSSA-N
57363
(1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
C23H36N2O2
MFCD00869737
finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
CHEBI:5062
CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
Specifications
98319-26-7
MFCD00869737
14,4082
Soluble in water,DMSO,ethanol,methanol,and chloroform.
CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
372.553
CHEBI:5062
Finasteride
C23H36N2O2
100mg
finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
DBEPLOCGEIEOCV-WSBQPABSSA-N
(1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
57363
372.54
Product Suggestions
Product Suggestions
Videos
SDS Documents