Learn More
Finasteride, Thermo Scientific™
Inhibitor of steroid 5-a-reductase
61.80€ - 810.00€
Chemical Identifiers
| CAS | 98319-26-7 |
|---|---|
| Molecular Formula | C23H36N2O2 |
| Molecular Weight (g/mol) | 372.553 |
| MDL Number | MFCD00869737 |
| InChI Key | DBEPLOCGEIEOCV-WSBQPABSSA-N |
| Synonym | finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin |
| PubChem CID | 57363 |
| ChEBI | CHEBI:5062 |
| IUPAC Name | (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| SMILES | CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15455699
|
Thermo Scientific Alfa Aesar
J63454.MC |
100mg |
61.80€
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15465699
|
Thermo Scientific Alfa Aesar
J63454.ME |
500mg |
113.00€
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15435699
|
Thermo Scientific Alfa Aesar
J63454.03 |
1g |
203.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15445699
|
Thermo Scientific Alfa Aesar
J63454.06 |
5g |
810.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsFinasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 98319-26-7 | |
| 372.553 | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| 57363 | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| C23H36N2O2 | |
| MFCD00869737 | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| CHEBI:5062 | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C |
Specifications
| 98319-26-7 | |
| MFCD00869737 | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide | |
| 57363 | |
| 372.54 |
| C23H36N2O2 | |
| 14,4082 | |
| Soluble in water,DMSO,ethanol,methanol,and chloroform. | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C | |
| 372.553 | |
| CHEBI:5062 | |
| Finasteride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H360D
missing translation for 'rtecsNumber' : CL5245000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C