missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filensin Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-46315JF669
This item is not returnable.
View return policy
Description
Filensin Polyclonal antibody specifically detects Filensin in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Filensin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD | |
| beaded filament structural protein 1, filensin. (Uniprot# Q12934) | |
| 0.1 mL | |
| Vision | |
| 631 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction