missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Rabbit anti-Human, Clone: FLG/1957R, Novus Biologicals™
Rabbit Monoclonal Antibody
551.00€
Specifications
| Antigen | Filaggrin |
|---|---|
| Clone | FLG/1957R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Filaggrin Monoclonal specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Filaggrin | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| Recombinant human Filaggrin protein fragment (around aa 198-288) (exact sequence is proprietary) | |
| Protein A or G purified |
| FLG/1957R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2312 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title