missing translation for 'onlineSavingsMsg'
Learn More

Fibulin-3/EFEMP1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

189.00€

Specifications

Antigen Fibulin-3/EFEMP1
Concentration 1 mg/mL
Dilution Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793264
View Documents
Novus Biologicals
NBP1-77040-0.025mg
0.025 mg
189.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fibulin-3/EFEMP1 Polyclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Fibulin-3/EFEMP1
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL
Polyclonal
Rabbit
Vision
PBS with 0.02% Sodium Azide
DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5
EFEMP1
IgG
Affinity Purified
At least three isoforms of Fibulin 3 are known to exist. This antibody is predicted to not cross-react with other Fibulin proteins.
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat
Q12805
2202
Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Fibulin 3. The immunogen is located within amino acids 410 - 460 of Fibulin 3. Amino Acid Squence: TFRIKSGNENGEFYLRQTS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
47 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.