missing translation for 'onlineSavingsMsg'
Learn More

Fibronectin Ab-11 Mouse Monoclonal Antibody, 200 μg/mL, BSA and azide,Lab Vision™ Epredia™

Product Code. 12609636 Shop All Epredia Products
Change view
Click to view available options
:
500μL; 200μg/mL; Unlabeled; Purified with BSA and azide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12609636 500μL; 200μg/mL; Unlabeled; Purified with BSA and azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12609636 Supplier Epredia Supplier No. MS1351P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunofluorescence experiments with Epredia™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: FBN11

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not known

Immunogen: Recombinant fibronectin protein

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin Ab-11
Applications Immunofluorescence
Classification Monoclonal
Clone FBN11
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant fibronectin protein
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.