missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma chain Antibody (OTI2D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45663
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Fibrinogen gamma chain | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:400-4000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
| Mouse | |
| 46.4 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 2266 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2D2 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| FGG | |
| Full length human recombinant protein of human FGG(NP_000500) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction